missing translation for 'onlineSavingsMsg'
Learn More

Succinylsulfathiazole, MedChemExpress

Product Code. 16418516
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16418516 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16418516 Supplier Medchem Express Supplier No. HYB0921
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Succinylsulfathiazole is a sulfonamide, it is an ultra long acting drug.

Succinylsulfathiazole is a sulfonamide, it is an ultra long acting drug.

Specifications

Chemical Name or Material Succinylsulfathiazole
CAS 116-43-8
Color White
Purity Grade Notes Research
Molecular Formula C13H13N3O5S2
Quantity 100 mg
Synonym Succinylsulphathiazole
Solubility Information DMSO : 100 mg/mL (281.38 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.81 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(O)CCC(NC1=CC=C(S(=O)(NC2=NC=CS2)=O)C=C1)=O
Molecular Weight (g/mol) 355.39
Formula Weight 355.39
Percent Purity 98.31%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.