Learn More
Sucrose Brix/RI Standards, Paragon Scientific™
Paragon ISO 17025 and ISO 17034 certified brix standards are for the calibration and verification of all types of refractometer e.g handheld, Abbe and high accuracy digital instruments. Typically used within the food and beverage and pharmaceutical industries.
Brand:  Paragon Scientific SS10
Additional Details : Weight : 0.01500kg
 View more versions of this product
Product Code. 16000410
  • 64.50 € / 15mL
Estimated Shipment: 17-05-2024
to see stock.
Description
Description
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • Primary level refractive index standards traceable to National Standards
  • Manufactured in accordance with ICUMSA Methods
  • Supplied in tamper evident packs with 6 weeks shelf life
Specifications
Specifications
10.00 Brix
15 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Nominal values quoted