missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sudan I, pure

Product Code. 10685382 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10685382

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 842-07-9 | C16H12N2O | 248.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 842-07-9
Molecular Formula C16H12N2O
Molecular Weight (g/mol) 248.285
MDL Number MFCD00003911
InChI Key ZONYAPYTDIVJGG-VLGSPTGOSA-N
Synonym C.I. 12055, 1-Phenylazo-2-naphthol, Solvent Yellow 14
PubChem CID 5858445
IUPAC Name (1Z)-1-(phenylhydrazinylidene)naphthalen-2-one
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC=CC=C32

Specifications

Melting Point 131.0°C to 133.0°C
Beilstein 16, IV, 228
Infrared Spectrum Authentic
Lambda Max at approximately 418 and 476nm (in ethanol)
Formula Weight 248.28
Grade Pure
Packaging Glass bottle
Specific Extinction 500 min. (at 476nm, in ethanol)
Color Brown or Orange to Red
Quantity 100 g
Physical Form Powder
Chemical Name or Material Sudan I
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sudan I
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Carcinogenicity Category 2
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H413-May cause long lasting harmful effects to aquatic life.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.