missing translation for 'onlineSavingsMsg'
Learn More

Sulfamethazine, TRC

Product Code. 17982055
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17982055 100 mg 100mg
17981845 10 mg 10mg
17900815 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17982055 Supplier Toronto Research Chemicals Supplier No. TRCS699074100MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 57-68-1
Molecular Formula C12 H14 N4 O2 S
Molecular Weight (g/mol) 278.33
Synonym Sulfanilamide, N1-(4,6-dimethyl-2-pyrimidinyl)- (8CI), 2-(4-Aminobenzenesulfonamido)-4,6-dimethylpyrimidine, 2-(4-Aminobenzenesulfonylamino)-4,6-dimethylpyrimidine, 2-(p-Aminobenzenesulfonamido)-4,6-dimethylpyrimidine, 2-Sulfanilamido-4,6-dimethylpyrimidine, 4,6-Dimethyl-2-sulfanilamidopyrimidine, 4-Amino-N-(2,6-dimethyl-4-pyrimidinyl)benzenesulfonamide, 4-Amino-N-(4,6-dimethyl-2-pyrimidinyl)benzenesulfonamide, 4-Amino-N-(4,6-dimethylpyrimidin-2-yl)benzenesulfonamide, A 502, Azolmetazin, BN 2409, Calfspan, Calfspan Tablets, Cremomethazine, Diazil, DiazilSulfadine, Diazyl, Dimezathine, Dimidin R, Kelametazine, Mermeth, N-(4,6-Dimethyl-2-pyrimidyl)sulfanilamide, N1-(4,6-Dimethyl-2-pyrimidinyl)sulfanilamide, N1-(4,6-Dimethyl-2-pyrimidyl)sulfanilamide, NSC 67457, NSC 683529, Neasina, Neazina, Panazin, Pirmazin, S-Dimidine, Spanbolet, SulfaSURE SR Bolus, Sulfadimerazine, Sulfadimesin, Sulfadimesine, Sulfadimethyldiazine, Sulfadimethylpyrimidine, Sulfadimezin, Sulfadimezine, Sulfadimidin, Sulfadimidine, Sulfadine, Sulfamethazine, Sulfamethiazine, Sulfodimesin, Sulfodimezine, Sulka K Boluses, Sulphadimethylpyrimidine, Sulphadimidine, Sulphamethasine, Sulphamethazine, Sulphamezathine, Sulphamidine, Sulphodimezine, Superseptil, Superseptyl, Sustain III, Vertolan, Benzenesulfonamide, 4-amino-N-(4,6-dimethyl-2-pyrimidinyl)-
IUPAC Name 4-amino-N-(4,6-dimethylpyrimidin-2-yl)benzenesulfonamide
SMILES Cc1cc(C)nc(NS(=O)(=O)c2ccc(N)cc2)n1

Specifica

Quantity 100 mg
InChI Formula InChI=1S/C12H14N4O2S/c1-8-7-9(2)15-12(14-8)16-19(17,18)11-5-3-10(13)4-6-11/h3-7H,13H2,1-2H3,(H,14,15,16)
Formula Weight 278.0837
Chemical Name or Material Sulfadimidine
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.