missing translation for 'onlineSavingsMsg'
Learn More

Sulfamonomethoxine, MedChemExpress

Product Code. 16494507
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494507 50 mg 50mg
16488638 10 mg 10mg
16428536 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16494507 Supplier Medchem Express Supplier No. HYB0946
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sulfamonomethoxine is a long acting sulfonamide antibacterial agent, used in blood kinetic studies,and blocks the synthesis of folic acid by inhibiting synthetase of dihydropteroate.

Sulfamonomethoxine is a long acting sulfonamide antibacterial agent, used in blood kinetic studies,and blocks the synthesis of folic acid by inhibiting synthetase of dihydropteroate.

Specifications

Chemical Name or Material Sulfamonomethoxine
Physical Form Solid
CAS 1220-83-3
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C11H12N4O3S
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 100 mg/mL (356.76 mM; Need ultrasonic) ∣H2O : 1 mg/mL (3.57 mM; ultrasonic and warming and heat to 80°C)
SMILES O=S(C1=CC=C(N)C=C1)(NC2=NC=NC(OC)=C2)=O
Molecular Weight (g/mol) 280.3
Formula Weight 280.3
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.