missing translation for 'onlineSavingsMsg'
Learn More

Sulfasalazine, MedChemExpress

Product Code. 16491429
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491429 1 g 1g
16448727 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16491429 Supplier Medchem Express Supplier No. HY14655
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sulfasalazine (NSC 667219) is an anti-rheumatic agent for the research of rheumatoid arthritis and ulcerative colitis. Sulfasalazine can suppress NF-κB activity. Sulfasalazine is a type 1 ferroptosis inducer.

Sulfasalazine (NSC 667219) is an anti-rheumatic agent for the research of rheumatoid arthritis and ulcerative colitis. Sulfasalazine can suppress NF-κB activity. Sulfasalazine is a type 1 ferroptosis inducer.

Especificaciones

Chemical Name or Material Sulfasalazine
CAS 599-79-1
Color Orange
Purity Grade Notes Research
Molecular Formula C18H14N4O5S
Quantity 1 g
Synonym NSC 667219
Solubility Information NH4OH : 150 mg/mL (376.52 mM; ultrasonic and adjust pH to 9 with NH4OH) ∣DMSO : 80 mg/mL (200.81 mM; Need ultrasonic and warming)
SMILES OC(C1=C(O)C=CC(/N=N/C2=CC=C(S(=O)(NC3=NC=CC=C3)=O)C=C2)=C1)=O
Molecular Weight (g/mol) 398.39
Formula Weight 398.39
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Mostrar más Mostrar menos

Research purposes only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.