missing translation for 'onlineSavingsMsg'
Learn More

Sulfate, Ion chromatography standard solution, Specpure™, SO4-2 1000μg/mL

Product Code. 11465010
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465010 500 mL 500mL
11353647 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11465010 Supplier Thermo Scientific Alfa Aesar Supplier No. 035547.AP
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sulfate, Ion chromatography standard solution is used as a standard solution in analytical chemistry and in ion chromatography. It is also used for calibration of ion chromatography and other analytical techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00145343

Specifications

Physical Form Liquid
Solubility Miscible with water
Type Analytical Standard
Quantity 500 mL
Grade Specpure
Chemical Name or Material Sulfate Ion chromatography standard solution, SO4-2 1000μg/mL

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.