missing translation for 'onlineSavingsMsg'
Learn More

Sulfisoxazole, TRC

Product Code. 30424238
Change view
Click to view available options
Quantity:
50 mg
1 g
100 g
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424238 50 mg 50mg
30425757 100 g -
30426156 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30424238 Supplier Toronto Research Chemicals Supplier No. TRCS68911050MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-69-5
Molecular Formula C11 H13 N3 O3 S
Molecular Weight (g/mol) 267.3
Synonym 3,4-Dimethyl-5-sulfanilamidoisoxazole, 3,4-Dimethyl-5-sulfonamidoisoxazole, 4-Amino-N-(3,4-dimethylisoxazol-5-yl)benzenesulfonamide, 5-(4-Aminophenylsulfonamido)-3,4-dimethylisoxazole, 5-(p-Aminobenzenesulfonamido)-3,4-dimethylisoxazole, 5-(p-Aminobenzenesulphonamido)-3,4-dimethylisoxazole, 5-Sulfanilamido-3,4-dimethylisoxazole, Alphazole, Amidoxal, Chemouag, Dorsulfan Warthausen, Entusul, Gantrisin, Gantrisona, Gantrizin, Gantrosan, Isoxamin, N-(3,4-Dimethylisoxazol-5-yl)-4-aminobenzenesulfonamide, N1-(3,4-Dimethyl-5-isoxazolyl)sulfanilamide, NSC 13120, NSC 33807, NSC 38588, NSC 683536, Neazolin, Neoxazol, Norilgan-S, Pancid, Roxosul tablets, Sosol, Soxisol, Soxomide, Stansin, Sulbio, Sulfadimethylisoxazole, Sulfafurazol, Sulfafurazole, Sulfagan, Sulfaisoxazole, Sulfalar, Sulfasol, Sulfazin, Sulfisoxazol, Sulfisoxazole, Sulfisoxazole dialamine, Sulfoxol, Sulphafurazole, Sulphisoxazole, Sulsoxin, Thiasin, Unisulf, Uritrisin
IUPAC Name 4-amino-N-(3,4-dimethyl-1,2-oxazol-5-yl)benzenesulfonamide
SMILES Cc1noc(NS(=O)(=O)c2ccc(N)cc2)c1C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H13N3O3S/c1-7-8(2)13-17-11(7)14-18(15,16)10-5-3-9(12)4-6-10/h3-6,14H,12H2,1-2H3
Formula Weight 267.0678
Chemical Name or Material Sulfisoxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.