missing translation for 'onlineSavingsMsg'
Learn More

Sulfosalicylic Acid, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15507834 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2 kg
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
2kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15507834 2 kg Wide Mouth Amber Glass Bottle 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15507834 Supplier Avantor J.T.Baker Supplier No. 036405
  • ONLINE EXCLUSIVE PRICE
    1680.00 €
    1508.12 € / 2kg
    Save 171.88 €
    10% Off
 Restricted Item
Estimated Shipment: 26-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 5965-83-3
Molecular Formula C9H14O6S
Molecular Weight (g/mol) 250.27
MDL Number MFCD00007508,MFCD00149540
InChI Key NFYHZVWMQHQKRU-UHFFFAOYSA-N
Synonym 2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd
PubChem CID 2723734
SMILES C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O

Specifications

Packaging Wide Mouth Amber Glass Bottle
Quantity 2 kg
IUPAC Name 2-hydroxy-5-sulfobenzoic acid;dihydrate
Formula Weight 254.22
Grade BAKER ANALYZED™ ACS Reagent
Physical Form Crystals
Chemical Name or Material Sulfosalicylic Acid, Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.