missing translation for 'onlineSavingsMsg'
Learn More

Sulfur @ 7000μg/g in Residual Oil, 500mL

Product Code. 30312858
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30312858 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312858 Supplier VHG Labs Supplier No. VHGSRES7000500
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfur
CAS 7704-34-9
Analyte or Component Names Sulfur
Shipping Condition Room Temperature
Molecular Formula S
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Synonym Sulfur, Schwefel, Agri-Sul, Alphagran S 80, Asulfa-Supra, Tiger 90 J
InChI Formula InChI=1 S/S
Solution Type Single Solution
SMILES [S]
Element S
IUPAC Name sulfur
Solvent or Matrix Residual Oil
Industry Type Petrochemical Testing
Molecular Weight (g/mol) 32.065
Quantity 500 mL
Formula Weight 31.972
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.