missing translation for 'onlineSavingsMsg'
Learn More

Sulfur in Crude Oil standard solution, 1000μg/g (0.10%), Specpure™

Product Code. 15426405
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15426405 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15426405 Supplier Thermo Scientific Alfa Aesar Supplier No. 041885.AE
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7704-34-9 | S | 32.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7704-34-9
Odor Unpleasant
Physical Form Liquid
Molecular Formula S
MDL Number MFCD00085316
Color Black
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
SMILES [S]
IUPAC Name sulfur
Molecular Weight (g/mol) 32.06
PubChem CID 5362487
Quantity 100 mL
ChEBI CHEBI:26833
Formula Weight 3350
Chemical Name or Material Sulfur, 1000μg/g (0.10%)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.