missing translation for 'onlineSavingsMsg'
Learn More

Sulfur-Free Calcium Concentrate: Ca @ approx. 2 wt% in Hydrocarbons, VHG™

Product Code. 30003106
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003106 100 g 100g
30002981 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30003106 Supplier VHG Labs Supplier No. VHGROSFCA2100G
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Sulfur-Free Calcium Concentrate: Ca @ approx. 2 wt% in Hydrocarbons
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium
CAS 7440-70-2
Methods XRF
Analyte or Component Names Calcium
Shipping Condition Room Temperature
Molecular Formula Ca
Synonym Ca
InChI Formula InChI=1S/Ca
SMILES [Ca]
IUPAC Name calcium
Solvent or Matrix Hydrocarbon Oil
Molecular Weight (g/mol) 40.078
Quantity 100 g
Formula Weight 39.963 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.