missing translation for 'onlineSavingsMsg'
Learn More

Sulfur-Free Magnesium Concentrate: Mg @ approx. 2 wt% in Hydrocarbons, VHG™

Product Code. 30003025
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 30003025

Brand: VHG Labs VHGROSFMG2100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Sulfur-Free Magnesium Concentrate: Mg @ approx. 2 wt% in Hydrocarbons
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zinc
CAS 7440-66-6
Methods XRF
Analyte or Component Names Zinc
Shipping Condition Room Temperature
Molecular Formula Zn
Synonym Zinc powder, Cinc, Merrillite
InChI Formula InChI=1S/Zn
SMILES [Zn]
IUPAC Name zinc
Solvent or Matrix Hydrocarbon Oil
Molecular Weight (g/mol) 65.41
Quantity 100 g
Formula Weight 63.9291 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.