missing translation for 'onlineSavingsMsg'
Learn More

Sulfur-Free Thallium Concentrate: Tl @ approx. 3 wt% in Hydrocarbons, VHG™

Product Code. 30003160
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003160 25 g 25g
30003037 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30003160 Supplier VHG Labs Supplier No. VHGROSFTL325G
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Sulfur-Free Thallium Concentrate: Tl @ approx. 3 wt% in Hydrocarbons
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thallium
CAS 7440-28-0
Methods XRF
Analyte or Component Names Thallium
Molecular Formula Tl
Synonym Ramor, Thallium, metallic, Thallium, elemental
InChI Formula InChI=1S/Tl
SMILES [Tl]
IUPAC Name thallium
Solvent or Matrix Hydrocarbon Oil
Molecular Weight (g/mol) 204.38
Quantity 25 g
Formula Weight 204.974 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.