missing translation for 'onlineSavingsMsg'
Learn More

Sulfur-Free Yttrium Concentrate: Y @ approx. 1 wt% in Hydrocarbons, VHG™

Product Code. 30003173
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003173 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30003173 Supplier VHG Labs Supplier No. VHGROSFY2100G
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Sulfur-Free Yttrium Concentrate: Y @ approx. 1 wt% in Hydrocarbons
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Yttrium
CAS 7440-65-5
Methods XRF
Analyte or Component Names Yttrium
Molecular Formula Y
Synonym Y
InChI Formula InChI=1S/Y
SMILES [Y]
IUPAC Name yttrium
Solvent or Matrix Hydrocarbon Oil
Molecular Weight (g/mol) 88.906
Quantity 100 g
Formula Weight 88.906 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.