missing translation for 'onlineSavingsMsg'
Learn More

Sulfur in Light Mineral Oil standard solution, Specpure™, 100μg/g (0.0100%)

Product Code. 15453855
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. Quantity unitSize
15453855 100 mL 100mL
1 options
This item is not returnable. View return policy

Product Code. 15453855

Brand: Thermo Scientific Alfa Aesar 042159.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

MDL Number MFCD00804270
Quantity 100 mL
Grade Specpure
Chemical Name or Material Sulfur in Light Mineral Oil standard solution, 100μg/g (0.0100%)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.