missing translation for 'onlineSavingsMsg'
Learn More

Sulfur Oil Standard in #2 Diesel Fuel, 25μg/g, SPEX CertiPrep™

Product Code. 15206326 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15206326 100 mL 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15206326 Supplier SPEX Certiprep Supplier No. SDFS25Y
  • ONLINE EXCLUSIVE PRICE
    137.39 €
    115.71 € / 100g
    Save 21.68 €
    15% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
S25μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging Triple-leached LDPE bottle
CAS 544-40-1, 68476-34-6
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 100.0, 0.0
Analyte or Component Names Sulphur
Molecular Formula C8H18S
MDL Number MFCD00009468
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
InChI Key HTIRHQRTDBPHNZ-UHFFFAOYSA-N
Type Certified Reference Material
SMILES CCCCSCCCC
Element Sulphur
IUPAC Name 1-(butylsulfanyl)butane
Solvent or Matrix #2 Diesel Fuel
Industry Type Petrochemical Testing
Molecular Weight (g/mol) 146.29
PubChem CID 11002
Quantity 100 mL
Concentration or Composition (by Analyte or Components) 25μg/g
Chemical Name or Material Sulfur Oil Standard in #2 Diesel Fuel, For Diesel Fuel Analysis
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sulfur Oil Standard in 2 Diesel Fuel
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Fuels, diesel, No 2

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.