missing translation for 'onlineSavingsMsg'
Learn More

Sulfur, plasma standard solution, Specpure™ S 1000μg/mL

Product Code. 11381399
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381399 100 mL 100mL
11391399 50 mL 50mL
11460350 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11381399 Supplier Thermo Scientific Alfa Aesar Supplier No. 013842.AE
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

(NH4)2SO4 in H2O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula (NH4)2SO4 in H2O
MDL Number MFCD00085316

Specifications

Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Ammonium sulfate: 0.413%; Water: 99.587%
Chemical Name or Material Sulfur, plasma standard solution

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.