missing translation for 'onlineSavingsMsg'
Learn More

Sulfur Standard: S @ 20 μg/g (0.0020 wt%) in #2 Diesel Fuel, VHG™

Product Code. 17971816
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 gal
Unit Size:
1 GAL
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17971816 1 gal 1 GAL
17912606 500 mL 500mL
17922606 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17971816 Supplier VHG Labs Supplier No. VHGSDSL201GAL
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Sulfur Standard: S @ 20 μg/g (0.0020 wt%) in #2 Diesel Fuel
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfur
CAS 7704-34-9
Methods XRF
Analyte or Component Names Sulfur
Shipping Condition Room Temperature
Molecular Formula S
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/S
SMILES [S]
IUPAC Name sulfur
Solvent or Matrix #2 Diesel Fuel
Molecular Weight (g/mol) 32.065
Quantity 1 gal
Formula Weight 31.972 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.