missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, Acculute Standard Volumetric Solution, Final Concentration 2.0N, Thermo Scientific Chemicals

Product Code. 11359299
Click to view available options
Quantity:
1 Unit
6 Units
Unit Size:
1 unit
6 units
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11359299

Brand: Thermo Scientific Alfa Aesar 088568.LQ

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Odor Acrid
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Color Colorless to Yellow
Quantity 1 Unit
Formula Weight 98.08
Concentration or Composition (by Analyte or Components) Sulfuric acid: 55%; Water: 45%
Chemical Name or Material Sulfuric acid
What are the instructions for dilution of 1 unit of Sulfuric acid, Acculute Standard Volumetric Solution, Final Concentration 2.0N (088568.LQ, 088568-LQ)?

The following is the dilution protocol:
- Transfer entire contents of 1 unit bottle into a 1L volumetric flask.
- Rinse bottle residue, the threads of the bottle, and inner bottle cap each with DI water, and transfer to the 1L flask.
- Repeat rinse three times. Bring the 1L flask to volume with DI water and mix well.
The final solution in the 1L flask has a concentration of 2.0N Sulfuric acid.

What is 1 unit of Sulfuric acid, Acculute Standard Volumetric Solution, Final Concentration 2.0N (088568.LQ, 088568-LQ)?

1 unit of Sulfuric acid, Acculute Standard Volumetric Solution, Final Concentration 2.0N, when diluted to 1 L, will yield a final concentration of 2.0N Sulfuric acid solution.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.