missing translation for 'onlineSavingsMsg'
Learn More

Sulindac, MedChemExpress

Product Code. 16478056
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478056 100 mg 100mg
16441996 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16478056 Supplier Medchem Express Supplier No. HYB0008
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sulindac (MK-231) is an orally active nonsteroidal anti-inflammatory agent.

Sulindac is used to reduce pain, swelling, and joint stiffness from arthritis. Sulindac is also used for the research of arthritis of the spine, gouty arthritis. Sulindac, as an immunomodulatory agent, can downregulate PD-L1 through the blockade of NF-κB signaling and modulates the response of pMMR colorectal cancer (CRC) to anti-PD-L1 immunotherapy, inhibits the development and progression of colorectal cancer CRC. Sulindac also inhibits TGF-β1- induced epithelial-mesenchymal transition (EMT) and suppresses lung cancer cell migration and invasion via downregulation of SIRT1.

Specifications

Chemical Name or Material Sulindac
CAS 38194-50-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H17FO3S
Quantity 100 mg
Synonym MK-231
Solubility Information DMSO : 100 mg/mL (280.58 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(CC(C1=C2C=CC(F)=C1)=C(/C2=C/C3=CC=C(C=C3)S(C)=O)C)O
Molecular Weight (g/mol) 356.41
Formula Weight 356.41
Percent Purity 99.34%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.