missing translation for 'onlineSavingsMsg'
Learn More

Sulphate Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™
BIOPHARMA

Product Code. 12922205 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12922205 100 mL 100mL
12932205 250 mL 250mL
12942205 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12922205 Supplier Fisher Chemical Supplier No. A 045 100
  • ONLINE EXCLUSIVE PRICE
    60.10 €
    51.16 € / 100mL
    Save 8.94 €
    14% Off
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Solution Contains H2O
Concentration 1 g/L
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.