missing translation for 'onlineSavingsMsg'
Learn More

Sulphide 1000 ppm Ion Selective Electrode Standard, Reagecon
BIOPHARMA

Product Code. 11728475 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11728475 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11728475 Supplier Reagecon Supplier No. ISES5
  • ONLINE EXCLUSIVE PRICE
    110.00 €
    104.47 € / 500mL
    Save 5.53 €
    5% Off
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Can be used at suitable concentrations to calibrate or control measurements that use ion selective electrodes (ISE) as the sensing technology.

  • Ion Selective Electrode Reference Standard, Ion Selective Electrode Reference Material
  • Certified
  • NIST traceable
  • Accurate to ± 0.5%
  • Safe, compatible with all types of ISE Electrodes
  • Can be used as a calibrant for other techniques
  • Can be used for method validation or instrument qualification
  • Aqueous matrix
  • Guaranteed stability throughout its entire shelf life, even after opening the bottle.

Specifications

Description Accuracy ± 0.5%
Packaging HDPE Bottle GL45
Analyte or Component Names Sulphide
Physical Form Liquid
Concentration 1000 ppm ±0.5%
Element Sulfide
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.