missing translation for 'onlineSavingsMsg'
Learn More

Sulphur for AAS 1g/L H2O matrix, Fisher Chemical™
BIOPHARMA

Product Code. 12903215 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12903215 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12903215 Supplier Fisher Chemical Supplier No. B 049 500
  • ONLINE EXCLUSIVE PRICE
    60.00 €
    46.17 € / 500mL
    Save 13.83 €
    23% Off
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Bottle
CAS 7704-34-9
Physical Form Liquid
Molecular Formula S
Concentration 1 g/L ±0.002 g/L
MDL Number MFCD00085316
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
Type Flame Single-element Standard Solution
SMILES [S]
IUPAC Name sulfur
Solvent or Matrix Water
Molecular Weight (g/mol) 32.06
PubChem CID 5362487
Quantity 500 mL
ChEBI CHEBI:26833
Chemical Name or Material Sulphur
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.