missing translation for 'onlineSavingsMsg'
Learn More

Sulphur in Hydrocarbons, Jet Aviation Fuel, 250mL (15.2 mg/kg), Paragon Scientific™

Product Code. 16030060
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16030060 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16030060 Supplier Paragon Scientific Supplier No. CRMUSUKR
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for Mercaptan Sulphur Content ISO 3012

  • Certified according to ISO 17034 under UKAS accreditation by means of a method specific inter-laboratory study.
  • Fully traceable to international standards.
  • Ensures good interlaboratory correlation
  • Supplied in tamper evident UN compliant security pack
  • Pack size 250 mL
  • 12 months shelf life

Specifications

UN Number UN 1223
Quantity 250 mL
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sulfur in Kerosene, ASTM D 3227; ISO 3012
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: kerosine

Nominal values quoted. Certified Values will vary from batch to batch.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.