missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUMO2/3 Antibody (SR1240), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00 € - 449.00 €
Specifications
| Antigen | SUMO2/3 |
|---|---|
| Clone | SR1240 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18636834
|
Novus Biologicals
NBP3-21701-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18682715
|
Novus Biologicals
NBP3-21701-50ul |
50 μg |
338.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SUMO2/3 Monoclonal antibody specifically detects SUMO2/3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
| SUMO2/3 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| HSMT3, small ubiquitin-like modifier 2, SMT3A, SMT3B, SMT3H2, SUMO2, SUMO3 | |
| A synthesized peptide derived from human SUMO2/3 (Uniprot #: P61956, P55854) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1240 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 6613 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title