Learn More
SUN1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 11 publications
Brand:  Novus Biologicals NBP1-87396
 View more versions of this product
Product Code. 18799063
  • 572.00 € / 0.10mL
Description
SUN1 Polyclonal specifically detects SUN1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Western Blot Western Blot reactivity reported in scientific literature (PMID: 25210889)., Simple Western 1:20, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunoprecipitation Use in immunoprecipitation reported in scientific literature (PMID: 25210889)., Immunohistochemistry-Paraffin 1:200-1:500
This antibody was developed against Recombinant Protein corresponding to amino acids:QLLPTVEHLQLELDQLKSELSSWRHVKTGCETVDAVQERVDVQVREMVKLLFSEDQQGGSLEQLLQRFSSQFVSKGDLQTMLRDLQLQILRNVTHHVSVTKQLPTSEAVVSAVSEAGASGITEAQARAIVNS
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
KIAA0810UNC84AFLJ12407, MGC176649, Protein unc-84 homolog A, Sad1 and UNC84 domain containing 1, Sad1 unc-84 domain protein 1, Sad1/unc-84 protein-like 1, SUN domain-containing protein 1, unc-84 homolog A, unc-84 homolog A (C. elegans)
0.1 mL