missing translation for 'onlineSavingsMsg'
Learn More

Sunifiram, MedChemExpress

Product Code. p-200101990
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16419867

Brand: Medchem Express HY17550

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sunifiram (DM-235) is a piperazine derived ampakine-like drug which has nootropic effects in animal studies with significantly higher potency than piracetam.

Sunifiram (DM-235) is a piperazine derived ampakine-like drug which has nootropic effects in animal studies with significantly higher potency than piracetam.

Specifications

Chemical Name or Material Sunifiram
Physical Form Solid
CAS 314728-85-3
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C14H18N2O2
For Use With (Application) Neuroscience-Neurodegeneration
Synonym DM-235
Solubility Information H2O : 50 mg/mL (203.00 mM; Need ultrasonic) ∣DMSO : ≥ 2.6 mg/mL (10.56 mM)
SMILES CCC(N1CCN(C(C2=CC=CC=C2)=O)CC1)=O
Molecular Weight (g/mol) 246.3
Formula Weight 246.3
Percent Purity 99.89%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.