Learn More
3M™ Supplied Air Respirator System, 06972
Product Code. 31297052 Shop All 3M Products
Change view
Click to view available options
Material:
Polypropylene
Product Code. Material
31297052 Polypropylene
1 options
Product Code. 31297052 Supplier 3M™ Supplier No. 7100361625
  • ONLINE EXCLUSIVE PRICE
    1150.00 €
    732.75 € / Pack of 1
    Save 417.25 €
    36% Off
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Provides respiratory protection against particles, gases and vapours, and combination hazards when used with an approved 3M face mask.
  • Compatible with 3M™ Full Facepiece 6000 Series and Half Face Mask 7500 Series.
  • Protects against particles, gases, vapours, and combination hazards.
  • Delivers constant airflow for enhanced comfort.
  • Double-action couplings help prevent accidental disconnection.
  • Adjustable airflow with Flow Check Bag for pre-use testing.
  • Low-profile breathing tube allows free movement and minimal interference.
  • Belt-mounted design with stainless-steel connection for durability and corrosion resistance.
  • ATEX-tested for safe use in IIB 2GD environments.
  • CE approved and compliant with EN14594:2005 standards.
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Product Type Supplied Air Regulator
For Use With (Application) Cleaning, Electrical, Facility Maintenance, Painting, Sanding, Welding
For Use With (Equipment) 3M Face Masks
Model S200+