Learn More
3M™ Supplied Air Respirator System, S-200+
Product Code. 31297051 Shop All 3M Products
Change view
Click to view available options
Material:
Polypropylene
Product Code. Material
31297051 Polypropylene
1 options
Product Code. 31297051 Supplier 3M™ Supplier No. 7100360886
  • ONLINE EXCLUSIVE PRICE
    1075.00 €
    686.02 € / Pack of 1
    Save 388.98 €
    36% Off
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Provides respiratory protection against particles, gases and vapours, and combination hazards when used with an approved 3M face mask.
  • Provides protection against fine dusts, aerosols, paint spray, metal fume, gases, vapours, and combination hazards.
  • Designed for use with approved 3M™ Half and Full Face Masks.
  • Can be used as a standard airline respirator or in dual-mode with filter back-up when the air supply is disconnected.
  • Lightweight, balanced design with a constant flow of user-adjustable air for cooler, more comfortable wear.
  • Low-profile, narrow-bore breathing tube allows easy movement and reduces work interference.
  • Belt-mounted system with double-action couplings to minimise the risk of accidental disconnection.
  • Stainless-steel CAST connection improves corrosion resistance for tough working conditions.
  • Includes a flow check bag for simple pre-use airflow verification.
  • Easy to maintain with quick and intuitive part replacement.
  • ATEX tested and suitable for use in IIB 2GD environments under specified conditions.
  • Tested in accordance with EN-13463-1 for non-electrical equipment when used with 3M™ 308-00-72P Antistatic Compressed Air Supply Tube.
  • CE approved and rated to EN14594:2005 Class 3A/B (half mask) or Class 4A/B (full face mask).
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Product Type Supplied Air Regulator
For Use With (Application) Cleaning, Electrical, Facility Maintenance, Painting, Sanding, Welding
For Use With (Equipment) 3M Face Masks
Model S200+