Learn More
Thermo Scientific™ SureSTART™ WebSeal™ 96-Well Microtiter Plates, Total V-Bottom, Level 2 High-Throughput Applications
Product Code. 17759598 Shop All Thermo Scientific Products
Change view
Click to view available options
Array:
8 x 12
Unit Size:
Pack of 50
Product Code. Array Unit Size
17759598 8 x 12 Pack of 50
1 options
Product Code. 17759598 Supplier Thermo Scientific™ Supplier No. 60180P210B
Estimated Shipment: 27-11-2026
Add to basket
Edge
Add to basket
Bulk Quote Request
missing translation for 'validMainframeMsg'
Use SureSTART™ WebSeal™ 96-Well Microtiter Plates, Total V-Bottom, Performance Level 2, for high-throughput analysis. They are certified and conform to ANSI-SLAS 4-2004 standards.
Thermo Scientific™ SureSTART™ Level 2 WebSeal™ 96-Well Total V-Bottom Microtiter Plates are designed to ensure high quality data with an uninterrupted workflow for high-throughput GC and HPLC/UHPLC applications. They are ideal for regulated labs that use water and polar solvents.
  • Performance Level 2 Plates for High Throughput Applications
  • Ideal for non-regulated labs that use water and polar solvents
  • Certified plastic well plates, seals, tapes
  • GC-MS and LC-MS tested
  • Suitable for HPLC/UHPLC applications
  • Suitable for QA/QC workflows and sensitive applications with low sample concentrations
  • Ideal for verifying compound presence and quantification using detection techniques such as UV, DAD/PDA, RI, Fluorescence, and ELSD
  • Specification Certified
  • All Thermo Scientific SureSTART WebSeal well plates are dimensionally verified
TRUSTED_SUSTAINABILITY
Specifications
Barcode Includes Barcode
Format SBS Standard
Array 8 x 12
Product Line SureSTART WebSeal Level 2
Color Clear
Product Type 96-Well Microtiter Plate
Material Polyethylene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-coated
Well Shape Round