missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SURF4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94778-0.02ml
This item is not returnable.
View return policy
Description
SURF4 Polyclonal antibody specifically detects SURF4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SURF4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin | |
| ERV29, FLJ22993, MGC102753, SURF-4, surface 4 integral membrane protein, surfeit 4, surfeit locus protein 4 | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human SURF4 (NP_001267719.1). ACFGLFGIIALQTIAYSILWDLKFLMRNLALGGGLLLLLAESRSEGKSMFAGVPTMRESSPKQYMQLGGRVLLVLMFMTLLHFDASFFSIVQNIVGTALMI | |
| 0.02 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6836 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction