missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SURF5 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93744-0.02ml
This item is not returnable.
View return policy
Description
SURF5 Polyclonal antibody specifically detects SURF5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SURF5 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| Med24, mediator complex subunit 22SURF5surfeit 5, mediator of RNA polymerase II transcription subunit 22, MGC48682, Surf-5, Surfeit locus protein 5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human MED22 (NP_852468.1). MAQQRALPQSKETLLQSYNKRLKDDIKSIMDNFTEIIKTAKIEDETQVSRATQGEQDNYEMHVRAANIVRAGESLMKLVSDLKQFLILNDFPSVNEAIDQRNQQLRTLQEECDRKLITLRDEISIDLYELEEEYYSSRYK | |
| 0.02 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6837 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction