missing translation for 'onlineSavingsMsg'
Learn More

Surrogate Standard, o-Terphenyl for Diesel Gasoline, SPEX CertiPrep™

Product Code. 16389087 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Dimensione della confezione
16389087 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 16389087 Fornitore SPEX Certiprep N. del fornitore DIESELI

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
o-Terphenyl84-15-15000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifica

Packaging 2mL amber prescored glass ampule
CAS 84-15-1, 75-09-2
CAS Min % 0.5, 99.5
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.5, 99.5
Physical Form Fluid
Molecular Formula C18H14
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key OIAQMFOKAXHPNH-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C2=CC=CC=C2C3=CC=CC=C3
IUPAC Name 1,2-diphenylbenzene
Solvent or Matrix Methylene Chloride
Molecular Weight (g/mol) 230.31
PubChem CID 6766
Quantity 1 mL
Chemical Name or Material o-Terphenyl
Vedi altri risultati Mostra meno risultati

Product intended for laboratory use only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.