missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Survivin, p Ser20 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
218.00 € - 450.00 €
Specifications
| Antigen | Survivin (p Ser20) |
|---|---|
| Concentration | 1.25 mg/mL |
| Dilution | Western Blot reported in scientific literature (PMID 24069188), Immunohistochemistry 1:10-1:500. Use reported in scientific literature (PMID 22814318), Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18169669
|
Novus Biologicals
NB110-92717SS |
0.025 mL |
218.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18706963
|
Novus Biologicals
NB110-92717 |
0.1 mL |
450.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Survivin Polyclonal specifically detects Survivin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| Survivin (p Ser20) | |
| Western Blot reported in scientific literature (PMID 24069188), Immunohistochemistry 1:10-1:500. Use reported in scientific literature (PMID 22814318), Immunocytochemistry/Immunofluorescence 1:100 | |
| Polyclonal | |
| Rabbit | |
| Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Phospho Specific, Stem Cells | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha | |
| BIRC5 | |
| IgG | |
| Affinity Purified | |
| 16 kDa |
| 1.25 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Rat | |
| O15392 | |
| 332 | |
| This Survivin [p Ser20] Antibody was developed against a synthetic peptide surrounding the phosphorylated serine 20 of the human Survivin protein. [Swiss-Prot #O15392] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title