missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUV420H2 Antibody (RAB-C409) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
244.00 € - 578.00 €
Specifications
| Antigen | SUV420H2 |
|---|---|
| Clone | RAB-C409 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Mass Spectrometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18684714
|
Novus Biologicals
NBP3-20151-0.025mg |
0.025 mg |
244.00 €
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18648774
|
Novus Biologicals
NBP3-20151-0.2mg |
0.2 mg |
578.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SUV420H2 Monoclonal antibody specifically detects SUV420H2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP assay (ChIP)Specifications
| SUV420H2 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| 84787 | |
| IgG κ | |
| Protein A purified |
| RAB-C409 | |
| Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Mass Spectrometry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| EC 2.1.1.43, FLJ98627, histone-lysine N-methyltransferase SUV420H2, KMT5Csuv4-20h2, Lysine N-methyltransferase 5C, MGC2705, Su(var)4-20 homolog 2, Suppressor of variegation 4-20 homolog 2, suppressor of variegation 4-20 homolog 2 (Drosophila), Suv4-20h2 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing SUV420H2 protein under non-denaturing conditions. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title