Learn More
Eppendorf™ Swing Bucket Rotors
Perfect for phase-separation applications such as phenol-chloroform extraction
Brand:  Eppendorf™ 5409715003
Code : A0
Additional Details : Weight : 1.80000kg
 View more versions of this product
Product Code. 13413406
  • 1330.00 € / Each
In Stock
to see stock.
Description
Description
  • Designed for use with Eppendorf Centrifuges
Specifications
Specifications
Swinging Bucket Rotor
16,049 x g
5427R Centrifuge
24 x 1.5/2 mL
12,700 rpm
QuickLock rotor lid, wind shield