Learn More
Eppendorf™ Swing Bucket Rotors
Perfect for phase-separation applications such as phenol-chloroform extraction
1330.00 €
Specifications
Type | Swinging Bucket Rotor |
---|---|
Capacity | 24 x 1.5/2 mL |
Max. RCF | 16,049 x g |
Max. Speed | 12,700 rpm |
Product Code | Brand | Capacity | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Includes | Price | Quantity & Availability | ||||
13413406
|
Eppendorf™
5409715003 |
24 x 1.5/2 mL | QuickLock rotor lid, wind shield |
1330.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Designed for use with Eppendorf Centrifuges
Specifications
Swinging Bucket Rotor | |
24 x 1.5/2 mL | |
16,049 x g | |
12,700 rpm |