missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
316.00 € - 456.00 €
Specifications
| Antigen | SYK |
|---|---|
| Concentration | 0.4mg/mL |
| Dilution | Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18489850
|
Novus Biologicals
NBP1-86178-25ul |
25 μL |
316.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18228017
|
Novus Biologicals
NBP1-86178 |
0.1 mL |
456.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SYK Polyclonal specifically detects SYK in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SYK | |
| Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Unconjugated | |
| RUO | |
| Human | |
| DKFZp313N1010, EC 2.7.10, EC 2.7.10.2, FLJ25043, spleen tyrosine kinaseFLJ37489, tyrosine-protein kinase SYK | |
| SYK | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| 0.4mg/mL | |
| Polyclonal | |
| Rabbit | |
| Immunology, Innate Immunity, Mast Cell Markers, Protein Kinase | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 6850 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GLLRVLTVPCQKIGTQGNVNFGGRPQLPGSHPATWSAGGIISRIKSYSFPKPGHRKSSPAQGNRQESTVSFNPYEPELAPWAADKGPQREALPMDTEVYESPYADPEEIRPKEVYLDRKLLTLEDKELGSGNF | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title