missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Syndecan 3 Monoclonal Antibody (312607)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561200
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
The protein encoded by this gene belongs to the syndecan proteoglycan family. It may play a role in the organization of cell shape by affecting the actin cytoskeleton, possibly by transferring signals from the cell surface in a sugar-dependent mechanism. Allelic variants of this gene have been associated with obesity.
Specifications
| Syndecan 3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q64519 | |
| Sdc3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Syndecan-3, Ala45-Glu384. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 312607 | |
| Unconjugated | |
| Sdc3 | |
| Kiaa0468; mKIAA0468; Neuroglycan; N-syndecan; SDC3; SDCN; syn-3; Synd3; syndecan 1; syndecan 3; syndecan neural type; syndecan proteoglycan 3; syndecan-3; wu:fa97a01 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 20970 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction