missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syndecan-4 Rabbit anti-Mouse, Clone: 112, Novus Biologicals™
Rabbit Monoclonal Antibody
392.00 € - 578.00 €
Specifications
| Antigen | Syndecan-4 |
|---|---|
| Clone | 112 |
| Dilution | Western Blot 1:500-1:1000, Flow Cytometry 1:25-1:100 |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18201188
|
Novus Biologicals
NBP2-90701-50UL |
50 μg |
392.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18206956
|
Novus Biologicals
NBP2-90701-100UL |
100 μg |
578.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Syndecan-4 Monoclonal specifically detects Syndecan-4 in Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
| Syndecan-4 | |
| Western Blot 1:500-1:1000, Flow Cytometry 1:25-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 6385 | |
| IgG | |
| Protein A purified |
| 112 | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| amphiglycan, MGC22217, ryudocan, Ryudocan core protein, SYND4ryudocan amphiglycan, syndecan 4, syndecan 4 (amphiglycan, ryudocan), syndecan proteoglycan 4, syndecan-4 | |
| Recombinant Mouse Syndecan-4 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title