missing translation for 'onlineSavingsMsg'
Learn More

Synephrine hydrochloride, MedChemExpress

Product Code. 16448996
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448996 100 mg 100mg
16402356 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16448996 Supplier Medchem Express Supplier No. HYN0132A
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Synephrine (Oxedrine) hydrochloride, an alkaloid, is an α-adrenergic and β-adrenergic agonist derived from the Citrus aurantium. Synephrine hydrochloride is a sympathomimetic compound and can be used for weight loss.

Synephrine (Oxedrine) hydrochloride, an alkaloid, is an α-adrenergic and β-adrenergic agonist derived from the Citrus aurantium. Synephrine hydrochloride is a sympathomimetic compound and can be used for weight loss.

Specifications

Chemical Name or Material Synephrine hydrochloride
CAS 5985-28-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C9H14ClNO2
Quantity 100 mg
Synonym Oxedrine hydrochloride
Solubility Information H2O : ≥ 100 mg/mL (490.99 mM) ∣DMSO : ≥ 52 mg/mL (255.31 mM)
SMILES OC(C1=CC=C(O)C=C1)CNC.Cl
Molecular Weight (g/mol) 203.67
Formula Weight 203.67
Percent Purity 99.57%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.