missing translation for 'onlineSavingsMsg'
Learn More

SynGAP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

213.00 € - 507.00 €

Specifications

Antigen SynGAP
Concentration 1 mg/mL
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/ Immunofluorescence 1 μg/mL, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Applications Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18536804
View Documents
Novus Biologicals
NB120-3344-0.025mg
0.025 mg
213.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18544630
View Documents
Novus Biologicals
NB120-3344
0.1 mg
507.00 €
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SynGAP Polyclonal antibody specifically detects SynGAP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications

Specifications

SynGAP
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/ Immunofluorescence 1 μg/mL, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Polyclonal
Purified
RUO
Human, Mouse, Rat
DKFZp761G1421, KIAA1938135kDa, MRD5, synaptic Ras GTPase activating protein 1, synaptic Ras GTPase activating protein 1 homolog (rat), Synaptic Ras-GAP 1
GST fusion protein of a recombinant fragment, residues 947-1167, of rat SynGAP.
Primary
Store at -20°C. Avoid freeze-thaw cycles.
1 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Neuroscience, Vision
PBS with 1 mg/mL BSA
8831
IgG
Immunogen affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.