missing translation for 'onlineSavingsMsg'
Learn More

Synthetic Sea Water for ASTM 665 / IP 135, Paragon Scientific™

Product Code. 16070420
Change view
Click to view available options
Quantity:
300 mL
500 mL
10 x 30 mL
5 L
Unit Size:
300mL
5 L
500mL
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16070420 10 x 30 mL Pack of 10
16080420 300 mL 300mL
16090420 500 mL 500mL
16000430 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16070420 Supplier Paragon Scientific Supplier No. SSWS30
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

A high quality analytical reagent manufactured and certified compliant for use in ASTM D 665 / IP 135 the Standard Test Method for Rust-Preventing Characteristics of Inhibited Mineral Oil in the Presence of Water.

  • A Synthetic Sea Water Analytical Reagent manufactured in accordance with ASTM D 665 / IP 135.
  • Provided with a certificate which includes an estimation of uncertainty of manufacture.
  • Ensures full compliance with ASTM and IP test method requirements for reagent preparation.
  • Conveniently packaged in single dose or multi dose packs.
  • Cost effective and time saving
  • Certificate of Manufacture & MSDS included
  • 12 month shelf life

Specifications

Certifications/Compliance ASTM 665, IP 135
Quantity 10 x 30 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.