missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synuclein alpha/beta Antibody (SR1281), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00 € - 449.00 €
Specifications
| Antigen | Synuclein alpha/beta |
|---|---|
| Clone | SR1281 |
| Dilution | Western Blot 1:1000-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18631794
|
Novus Biologicals
NBP3-21662-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18602474
|
Novus Biologicals
NBP3-21662-50ul |
50 μg |
338.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Synuclein alpha/beta Monoclonal antibody specifically detects Synuclein alpha/beta in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
| Synuclein alpha/beta | |
| Western Blot 1:1000-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| beta-synuclein, I(2)-synuclein, I+/--synuclein, NACP, non A-beta component of AD amyloid, PARK1, PD1, synuclein alpha-140, synuclein, alpha (non A4 component of amyloid precursor), truncated alpha synuclein | |
| A synthesized peptide derived from human Synuclein alpha/beta (Uniprot #: P37840, Q16143) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1281 | |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 6622 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title