missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYPL1 Recombinant Rabbit Monoclonal Antibody (K01_1Y98)

Product Code. 30928573
Change view
Click to view available options
Quantity:
50 μL
100 μL
Unit Size:
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30928573 50 μL 50µL
30825416 100 μL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30928573 Supplier Invitrogen™ Supplier No. MA564966

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: IKIATGHNII DELPPCKKKA VLCYFGSVTS MGSLNVSVIF GFLNMILWGG NAWFVYKETS LHSPSNTSAP HSQGGIPPPT GI Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 41%, Pig - 82%, Cynomolgus monkey - 94%.

The synaptophysin-like protein 1, also known as pantophysin, is a homolog of the neuroendocrine-specific protein synaptophysin, with the highest level of homology across its four transmembrane domains. Unlike synaptophysin however, SYPL1 is ubiquitously expressed and found in small cytoplasmic transport vesicles regardless of their content. SYPL1 is thought to play a multifunctional role in vesicle biogenesis and transport and is a component of adipocyte transport vesicles, and thus may be involved in adipocyte secretion. SYPL1 also interacts with vesicle-associated membrane protein 2 (VAMP-2) in adipocytes and associates with GLUT4-containing vesicles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYPL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_1Y98
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene SYPL1
Gene Accession No. Q16563
Gene Alias AI314763; AI604763; D12Ertd446e; H-SP1; Pan I; PanI; Pantophysin; Pphn; synaptophysin like 1; synaptophysin-like 1; synaptophysin-like protein; synaptophysin-like protein 1; SYPL; Sypl1
Gene Symbols SYPL1
Host Species Rabbit
Immunogen Recombinant protein of human SYPL1 (aa 178-259).
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6856
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.