Learn More
SYT8 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20671-1-AP-20UL
This item is not returnable.
View return policy
Description
The SYT8 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human SYT8. This antibody recognizes human, mouse antigen. The SYT8 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| SYT8 | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, Immunofluorescence, ELISA | |
| Synaptotagmin 8, synaptotagmin VIII, SYT8, SytVIII | |
| 20 μL | |
| 90019 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |