missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ SYTOX™ Green Dead Cell Stain, for flow cytometry

Product Code. 11310972
Change view
Click to view available options
Quantity:
1 mL, 1000 assays
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11310972 1 mL, 1000 assays Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11310972 Supplier Molecular Probes™ Supplier No. S34860
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

SYTOX™ Green Dead Cell Stain is a bright, easy-to-use nucleic acid stain for distinguishing dead from live cells in flow cytometry assays.

  • A bright, easy-to-use nucleic acid stain for distinguishing dead from live cells in flow cytometry assays
  • Designed for flow cytometry using the 488 nm blue laser with emission in the 530/30 BP filter (similar to FITC and GFP)
  • The dye concentration has been optimized for samples with concentrations ranging from 1 x 104 to 1 x 106 cells/mL
  • Stain has been validated in several cell types including Jurkat, ReH, CHO, Kg1a, 3T3, HeLa, HEK (primary cells), and lysed whole blood
  • Tested in several applications including apoptosis, cell cycle, cell proliferation, and immunophenotyping assays
TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Impermeant
Color Green
Content And Storage Contains 1 vial of SYTOX™ green dead cell stain (30 μM solution in DMSO).

Store in freezer (-5°C to -30°C), and protect from light.
Description SYTOX™ Green Dead Cell Stain, for flow cytometry
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Dead Cell Stain
Dye Type SYTOX™ Green
Emission 525
Excitation Wavelength Range 488 nm
Form Solution
Format Tube(s)
Product Line SYTOX
Quantity 1 mL, 1000 assays
Shipping Condition Room Temperature
Solubility DMSO (Dimethylsulfoxide)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.