missing translation for 'onlineSavingsMsg'
Learn More

T-26c, MedChemExpress

Product Code. 16472596
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16472596 25 mg 25mg
16463058 10 mg 10mg
16419626 50 mg 50mg
16415797 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16472596 Supplier Medchem Express Supplier No. HY100518
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress T-26c is highly potent and selective matrix metalloproteinase-13 (MMP-13) inhibitor with an IC50 of 6.75 pM and more than 2600-fold selectivity over the other related metalloenzymes.

T-26c is highly potent and selective matrix metalloproteinase-13 (MMP-13) inhibitor with an IC50 of 6.75 pM and more than 2600-fold selectivity over the other related metalloenzymes.

Specifications

Chemical Name or Material T-26c
CAS 869296-13-9
Color White
Purity Grade Notes Research
Molecular Formula C24H21N3O6S
Quantity 25 mg
Solubility Information DMSO : 15.62 mg/mL (32.57 mM; Need ultrasonic)
SMILES COC1=CC=CC(CNC(C2=NC(SC=C3COCC4=CC=C(C(O)=O)C=C4)=C3C(N2)=O)=O)=C1
Molecular Weight (g/mol) 479.51
Formula Weight 479.51
Percent Purity 99.54%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.