missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ T-bet Recombinant Rabbit Monoclonal Antibody (8V7M8)

Product Code. 30283161
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30283161 100 μL 100µL
30284407 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30283161 Supplier Invitrogen™ Supplier No. MA558098
  • ONLINE EXCLUSIVE PRICE
    457.00 €
    393.02 € / 100µL
    Save 63.98 €
    14% Off
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: KGFRENFESM YTSVDTSIPS PPGPNCQFLG GDHYSPLLPN QYPVPSRFYP DLPGQAKDVV PQAYWLGAPR DHSYEAEFRA VSMKPAFLPS APGPTMSYYR GQEVLAPGAG WPVAPQYPPK MGPASWFRPM RTLPMEPGPG GSEGRGPEDQ GPPLVWTEIA PIRPESSDSG LGEGDSKRRR VSPYPSSGDS SSPAGAPSPF DKEAEGQFYN YFPN.

T-bet, also known as TBX21, is a T-cell specific transcription factor containing a single T-box DNA-binding domain. It controls the expression of the TH1 cytokine, interferon-gamma. Both Th1 and Th17 cells are crucial in immune regulation and autoimmune disease development. T-bet initiates Th1 lineage development from naive Th precursor cells and directs T cell differentiation to Th1 versus Th17. T-bet is cooperating with Stat4 (signal tranducer and activator of transcription 4) in programming chromatin architecture for Th1 gene expression. Genetic variations in T-bet are associated with susceptibility to aspirin-induced asthma and nasal polyps.
TRUSTED_SUSTAINABILITY

Specifications

Antigen T-bet
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 8V7M8
Concentration 1.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Tbx21
Gene Accession No. Q9UL17
Gene Alias HGNC:11599; LOW QUALITY PROTEIN: T-box transcription factor TBX21; Tbet; T-bet; TBLYM; T-box 21; T-box expressed in T cells; T-box protein 21; T-box transcription factor 21; T-box transcription factor TBX21; TBT1; TBX21; T-cell-specific T-box transcription factor T-bet; T-PET; transcription factor TBLYM; transcription factor T-box 21
Gene Symbols Tbx21
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 322-535 of human T-bet/Tbx21 (NP_037483.1).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30009
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.